KEY017861229 Key Organics / BIONET

6-(1H-pyrrol-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD00828638
  • Cas Number : 218157-81-4
  • Molecular Weight : 169.187

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101 - 103
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7232

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11W-0847_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart