KEY017861293 Key Organics / BIONET

1,3-dimethyl 2-[2-nitro-4-(piperidine-1-sulfonyl)phenyl]propanedioate

  • MDL number MFCD04124523
  • Cas Number : 860785-48-4
  • Molecular Weight : 400.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109 - 111
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9932

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart