KEY017861337 Key Organics / BIONET

2-methyl-3-oxo-2H,3H,4H-pyrido[3,2-b][1,4]oxazine-2-carboxylic acid

  • MDL number MFCD04124560
  • Cas Number : 154365-45-4
  • Molecular Weight : 208.173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181 - 183
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9976

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0251_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart