KEY017861525 Key Organics / BIONET

methyl 2-chloro-4-(1H-pyrazol-1-yl)benzoate

  • MDL number MFCD04124715
  • Cas Number : 313674-09-8
  • Molecular Weight : 236.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65979

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-1485_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart