KEY017861623 Key Organics / BIONET

1-(4-methoxyphenyl)-2-(4-methylbenzenesulfonyl)ethan-1-one

  • MDL number MFCD00220318
  • Cas Number : 86516-51-0
  • Molecular Weight : 304.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130 - 131
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66394

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0841_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart