KEY017861792 Key Organics / BIONET

methyl 3-[(1E)-(hydroxyimino)methyl]-1H-indole-2-carboxylate

  • MDL number MFCD04125006
  • Cas Number : 861211-41-8
  • Molecular Weight : 218.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206 - 208
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69907

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0817_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart