KEY017861794 Key Organics / BIONET

1-[(6-chloropyridin-3-yl)methyl]-4-(2,6-dimethylphenyl)piperazine

  • MDL number MFCD04125008
  • Cas Number : 861211-43-0
  • Molecular Weight : 315.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69909

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart