KEY017862415 Key Organics / BIONET

2-[2-(2-amino-1,3-thiazol-4-yl)ethyl]-2,3-dihydro-1H-isoindole-1,3-dione hydrobromide

  • MDL number MFCD04125720
  • Cas Number : 95914-09-3
  • Molecular Weight : 354.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 266 - 267
Iupac Name 2-[2-(2-amino-1,3-thiazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione hydrobromide
Inchi Code 1S/C13H11N3O2S.BrH/c14-13-15-8(7-19-13)5-6-16-11(17)9-3-1-2-4-10(9)12(16)18;/h1-4,7H,5-6H2,(H2,14,15);1H
Inchi Key IPOCGIVMVZXNBQ-UHFFFAOYSA-N

Description

Melting Point 266 - 267
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80995

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5X-0266_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart