KEY017862450 Key Organics / BIONET

(E)-N-[(4-tert-butylphenyl)methylidene]hydroxylamine

  • MDL number MFCD01595462
  • Cas Number : 180261-48-7
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 105 - 107
Iupac Name 4-tert-butylbenzaldehyde oxime
Inchi Code 1S/C11H15NO/c1-11(2,3)10-6-4-9(5-7-10)8-12-13/h4-7H,8H2,1-3H3
Inchi Key MBACWGPQFXFCPP-UHFFFAOYSA-N

Description

Melting Point 105 - 107
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81041

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5X-0832_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart