KEY017862653 Key Organics / BIONET

2-{[3-(trifluoromethyl)phenyl]methoxy}naphthalene-1-carbaldehyde

  • MDL number MFCD04126040
  • Cas Number : 866133-96-2
  • Molecular Weight : 330.306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112 - 113
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/87979

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7W-0811_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart