KEY017862839 Key Organics / BIONET

3-amino-1-[3-(trifluoromethyl)phenyl]urea

  • MDL number MFCD01869439
  • Cas Number : 448233-17-8
  • Molecular Weight : 219.167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92 - 98
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94106

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9W-0814_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart