KEY020077302 Key Organics / BIONET

5-(2-bromophenyl)-1,3-oxazole

  • MDL number MFCD05668985
  • Cas Number : 328270-70-8
  • Molecular Weight : 224.057

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94274

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9X-0886_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart