KEY023878387 Key Organics / BIONET

{2-[(4-chlorophenyl)sulfanyl]phenyl}methanol

  • MDL number MFCD04108002
  • Cas Number : 13459-59-1
  • Molecular Weight : 250.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34 - 37
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/1587

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10K-578S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart