KEY023878389 Key Organics / BIONET

ethyl 3-(4-chlorophenyl)-5-hydroxy-1,2,4-triazine-6-carboxylate

  • MDL number MFCD04117779
  • Cas Number : 36286-77-8
  • Molecular Weight : 279.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247 - 250
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2100

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10M-511S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart