KEY023878393 Key Organics / BIONET

ethyl 2,4-bis(trifluoromethyl)-4aH,10aH-imidazo[1,2-a]1,8-naphthyridine-8-carboxylate

  • MDL number MFCD28385260
  • Cas Number : 439094-91-4
  • Molecular Weight : 379.262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189 - 191
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2856

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0339_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart