KEY023878414 Key Organics / BIONET

methyl 2-(1,2-benzoxazol-3-yl)acetate

  • MDL number MFCD04117784
  • Cas Number : 59899-89-7
  • Molecular Weight : 191.186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7145

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11W-0280_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart