KEY023878415 Key Organics / BIONET

2-(1,2-benzoxazol-3-yl)acetohydrazide

  • MDL number MFCD04117785
  • Cas Number : 23008-70-0
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 174
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7159

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11W-0304_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart