KEY023878416 Key Organics / BIONET

methyl 2-[(4-formyl-1,3-dimethyl-1H-pyrazol-5-yl)sulfanyl]benzoate

  • MDL number MFCD04117786
  • Cas Number : 852700-50-6
  • Molecular Weight : 290.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 - 99
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7196

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11W-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart