KEY023878420 Key Organics / BIONET

2-[3-(trifluoromethyl)phenoxy]pyridine-3-carboxylic acid

  • MDL number MFCD01938784
  • Cas Number : 36701-89-0
  • Molecular Weight : 283.206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156 - 159
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9452

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12P-365S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart