KEY023878423 Key Organics / BIONET

methyl 2-amino-5-bromopyridine-3-carboxylate

  • MDL number MFCD04117787
  • Cas Number : 50735-34-7
  • Molecular Weight : 231.049

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143 - 146
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/62879

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart