KEY023878424 Key Organics / BIONET

(E)-N-[(2-chloropyridin-3-yl)methylidene]hydroxylamine

  • MDL number MFCD04117788
  • Cas Number : 215872-96-1
  • Molecular Weight : 156.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 167
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/62880

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12W-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart