KEY023878428 Key Organics / BIONET

ethyl 1-(3,4-dichlorophenyl)-5-(trichloromethyl)-1H-1,2,4-triazole-3-carboxylate

  • MDL number MFCD04117789
  • Cas Number : 854019-67-3
  • Molecular Weight : 403.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 113
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64208

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1J-512S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart