KEY023878435 Key Organics / BIONET

ethyl 2-(1-benzylpiperidin-4-ylidene)-2-cyanoacetate

  • MDL number MFCD01569592
  • Cas Number : 1463-52-1
  • Molecular Weight : 284.359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64 - 67
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65709

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1R-1507_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart