KEY023878437 Key Organics / BIONET

methyl 2-chloro-4-(pyrrolidin-1-yl)benzoate

  • MDL number MFCD03844850
  • Cas Number : 175153-38-5
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 133 - 135
Iupac Name methyl 2-chloro-4-(1-pyrrolidinyl)benzoate
Inchi Code 1S/C12H14ClNO2/c1-16-12(15)10-5-4-9(8-11(10)13)14-6-2-3-7-14/h4-5,8H,2-3,6-7H2,1H3
Inchi Key CSBLQYDWEPLAOP-UHFFFAOYSA-N

Description

Melting Point 133 - 135
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65976

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1T-1482_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart