KEY023878438 Key Organics / BIONET

1-[4-(thiophen-2-yl)pyrimidin-2-yl]-1H-pyrrole-2-carbaldehyde

  • MDL number MFCD03791169
  • Cas Number : 855308-68-8
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 147
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/65995

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1T-9014_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart