KEY023878440 Key Organics / BIONET

1-[5-chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]ethan-1-ol

  • MDL number MFCD04117790
  • Cas Number : 959578-12-2
  • Molecular Weight : 228.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66354

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0702_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart