KEY023878442 Key Organics / BIONET

ethyl 1-(2-methyl-1-oxopropan-2-yl)piperidine-4-carboxylate

  • MDL number MFCD05663786
  • Cas Number : 886360-88-9
  • Molecular Weight : 227.304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1X-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart