KEY023878443 Key Organics / BIONET

2-amino-1-(4-methanesulfonylphenyl)ethan-1-one hydrochloride

  • MDL number MFCD05663787
  • Cas Number : 102871-96-5
  • Molecular Weight : 249.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 300
Iupac Name 2-amino-1-[4-(methylsulfonyl)phenyl]ethanone hydrochloride
Inchi Code 1S/C9H11NO3S.ClH/c1-14(12,13)8-4-2-7(3-5-8)9(11)6-10;/h2-5H,6,10H2,1H3;1H
Inchi Key JQZLJYCNKBOYGK-UHFFFAOYSA-N

Description

Melting Point 300
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66358

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0714_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart