KEY023878444 Key Organics / BIONET

4-(4-methanesulfonylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02663888
  • Cas Number : 383131-95-1
  • Molecular Weight : 254.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 254 - 256
Iupac Name 4-[4-(methylsulfonyl)phenyl]-1,3-thiazol-2-amine
Inchi Code 1S/C10H10N2O2S2/c1-16(13,14)8-4-2-7(3-5-8)9-6-15-10(11)12-9/h2-6H,1H3,(H2,11,12)
Inchi Key VQEMDSRIOVZAOM-UHFFFAOYSA-N

Description

Melting Point 254 - 256
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66359

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/1X-0715_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart