KEY023878449 Key Organics / BIONET

(1H-1,3-benzodiazol-5-yl)thiourea

  • MDL number MFCD03791189
  • Cas Number : 862665-25-6
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214 - 216
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68610

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2P-076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart