KEY023878451 Key Organics / BIONET

3-(1H-pyrrol-1-yl)-1-benzothiophene-2-carbohydrazide

  • MDL number MFCD06200944
  • Cas Number : 107363-01-9
  • Molecular Weight : 257.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129 - 132
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2R-1303_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart