KEY023878453 Key Organics / BIONET

ethyl 4-methyl-2-(pyridin-3-yl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD01567252
  • Cas Number : 39091-00-4
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59 - 60
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-0259_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart