KEY023878461 Key Organics / BIONET

methyl 4'-nitro-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD04117797
  • Cas Number : 5730-75-6
  • Molecular Weight : 257.245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188 - 190
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69884

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart