KEY023878466 Key Organics / BIONET

4-(2-amino-1,3-thiazol-4-yl)-2,6-dimethylphenol

  • MDL number MFCD04117801
  • Cas Number : 863305-85-5
  • Molecular Weight : 220.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126 - 130
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69889

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2X-0717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart