KEY023878473 Key Organics / BIONET

6-(methoxymethyl)-2-methylpyrimidin-4-ol

  • MDL number MFCD00087858
  • Cas Number : 38249-50-2
  • Molecular Weight : 154.169

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 181
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71942

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-543S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart