KEY023878474 Key Organics / BIONET

6-(methoxymethyl)-2-sulfanylpyrimidin-4-ol

  • MDL number MFCD00052620
  • Cas Number : 175205-07-9
  • Molecular Weight : 172.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230 - 232
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71943

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3M-544S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart