KEY023878475 Key Organics / BIONET

1-methyl-2-(4-methylpiperazin-1-yl)-1H-indole-3-carbaldehyde

  • MDL number MFCD05663789
  • Cas Number : 886360-89-0
  • Molecular Weight : 257.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143 - 145
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72357

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3P-335S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart