KEY023878478 Key Organics / BIONET

tert-butyl 4-{4-[(Z)-N'-hydroxycarbamimidoyl]phenyl}piperazine-1-carboxylate

  • MDL number MFCD05021842
  • Cas Number : 1233243-62-3
  • Molecular Weight : 320.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 206 - 207
Iupac Name tert-butyl 4-{4-[(Z)-amino(hydroxyimino)methyl]phenyl}-1-piperazinecarboxylate
Inchi Code 1S/C16H24N4O3/c1-16(2,3)23-15(21)20-10-8-19(9-11-20)13-6-4-12(5-7-13)14(17)18-22/h4-7,14H,8-11,17H2,1-3H3
Inchi Key SSIVZUCNKBMULR-UHFFFAOYSA-N

Description

Melting Point 206 - 207
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73390

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0269_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart