KEY023878483 Key Organics / BIONET

methyl 2-(4-formyl-2-nitrophenoxy)benzoate

  • MDL number MFCD04117808
  • Cas Number : 886360-60-7
  • Molecular Weight : 301.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73661

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart