KEY023878484 Key Organics / BIONET

methyl 2-(4-aminophenoxy)benzoate

  • MDL number MFCD04117809
  • Cas Number : 212189-58-7
  • Molecular Weight : 243.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 75 - 78
Iupac Name methyl 2-(4-aminophenoxy)benzoate
Inchi Code 1S/C14H13NO3/c1-17-14(16)12-4-2-3-5-13(12)18-11-8-6-10(15)7-9-11/h2-9H,15H2,1H3
Inchi Key FXHKMQZPYDWTCS-UHFFFAOYSA-N

Description

Melting Point 75 - 78
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73663

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3X-0711_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart