KEY023878485 Key Organics / BIONET

2-(4-nitrophenoxy)benzoic acid

  • MDL number MFCD01002507
  • Cas Number : 6082-87-7
  • Molecular Weight : 259.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 157
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73665

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart