KEY023878489 Key Organics / BIONET

(Z)-2-[(4-chlorophenyl)sulfanyl]-N'-hydroxypyridine-3-carboximidamide

  • MDL number MFCD05021868
  • Cas Number : 886360-64-1
  • Molecular Weight : 279.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 192
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73671

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart