KEY023878492 Key Organics / BIONET

3-chloro-4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}aniline

  • MDL number MFCD04117811
  • Cas Number : 76471-06-2
  • Molecular Weight : 323.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90 - 92
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73677

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3X-0731_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart