KEY023878506 Key Organics / BIONET

tert-butyl 4-[(pyrimidin-2-yl)amino]piperidine-1-carboxylate

  • MDL number MFCD04117813
  • Cas Number : 301225-40-1
  • Molecular Weight : 278.356

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132 - 133
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76216

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-322S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart