KEY023878507 Key Organics / BIONET

methyl 2-(pyridine-4-amido)benzoate

  • MDL number MFCD01033591
  • Cas Number : 60658-02-8
  • Molecular Weight : 256.261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124 - 127
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76751

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1532_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart