KEY023878508 Key Organics / BIONET

ethyl 5-amino-1-(4-methylphenyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00973936
  • Molecular Weight : 245.282

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114 - 116
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77066

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0243_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart