KEY023878516 Key Organics / BIONET

3-ethoxy-4-[(5-nitropyridin-2-yl)oxy]benzaldehyde

  • MDL number MFCD03791225
  • Cas Number : 702646-74-0
  • Molecular Weight : 288.259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 120
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77135

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4W-0345_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart