KEY023878518 Key Organics / BIONET

6-(piperidin-1-yl)pyridine-3-carbaldehyde

  • MDL number MFCD04117814
  • Cas Number : 241816-11-5
  • Molecular Weight : 190.246

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37 - 40
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77309

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4X-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart