KEY023878527 Key Organics / BIONET

3-[4-(benzyloxy)phenyl]propan-1-ol

  • MDL number MFCD03791226
  • Cas Number : 61440-45-7
  • Molecular Weight : 242.318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58 - 61
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80781

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0218_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart