KEY023878530 Key Organics / BIONET

2-chloro-5-(propan-2-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD03791228
  • Cas Number : 886360-70-9
  • Molecular Weight : 205.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 209 - 212
Iupac Name 2-chloro-5-isopropyl-1,3-thiazole-4-carboxylic acid
Inchi Code 1S/C7H8ClNO2S/c1-3(2)5-4(6(10)11)9-7(8)12-5/h3H,1-2H3,(H,10,11)
Inchi Key JBGWUKMKNIMSOU-UHFFFAOYSA-N

Description

Melting Point 209 - 212
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80861

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0350_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart